Learn More
Description
Lambda DNA digested with PvuI, 0.7% agarose, 3 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'..C G A T▵C G...3'
3'..G C▵T A G C...5'
Conditions for 100% Activity
- 1X Buffer R:10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer
- PvuI is supplied in:10mM Tris-HCl (pH 7.5 at 25°C), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.2mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage
- After 50-fold overdigestion with PvuI, more than 90% of the DNA fragments can be ligated and more than 95% of these can be recut
Methylation Effects
- Dam: completely overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: completely overlaps —blocked
- EcoKI: never overlaps —no effect
- EcoBI: never overlaps —no effect
Digestion of Agarose-embedded DNA
- Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours
Compatible Ends
- Bsp119I, Bsu15I, Hin1I, Hin6I, HpaII, MaeII, MspI, NarI, Psp1406I, SsiI, XmiI
Specifications
Specifications
| Concentration | 10 U/μL |
| Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
| Enzyme | Pvu I |
| Compatible Buffer | 10X Buffer R |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | No |
| Quantity | 300 units |
| Product Type | Restriction Enzyme |
| Research Category | Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.