Learn More
Thermo Scientific™ pUC19 DNA/MspI (HpaII) Marker
Shop All Thermo Scientific Products
Description
pUC19 DNA/MspI (HpaII) Marker is recommended for sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels. pUC19 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation.
pUC19 DNA/MspI (HpaII) Marker, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading onto agarose and polyacrylamide gels.
- Sizing and approximate quantification of small DNA fragments
- Sharp bands
- Supplied with loading dye for sample DNA
Specifications
Specifications
| Concentration | 6X |
| Gel Type | Agarose |
| Product Type | DNA/MSPI (HPAII) Marker |
| Ready to Load | No |
| Gel Compatibility | Agarose Gels |
| Green Features | Sustainable packaging |
| Product Line | pUC19 DNA/MspI (HpaII) Marker |
| Quantity | 5 x 50 μg |