Learn More
Thermo Scientific™ pUC19 DNA/MspI (HpaII) Marker
GREENER_CHOICE
Product Code. 10374680 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 x 50 μg
50 μg
Unit Size:
50µg
Pack of 5
Product Code. 10374680
Brand: Thermo Scientific™ SM0221
Perform sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels.
pUC19 DNA/MspI (HpaII) Marker is recommended for sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels. pUC19 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation.
pUC19 DNA/MspI (HpaII) Marker, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading onto agarose and polyacrylamide gels.
pUC19 DNA is digested to completion with the appropriate Fermentas PureExtreme restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the marker's preparation. pUC19 DNA/MspI (HpaII) Marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction.
  • Sizing and approximate quantification of small DNA fragments
  • Sharp bands
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Concentration 6X
Gel Type Agarose
Product Type DNA/MSPI (HPAII) Marker
Ready to Load No
Gel Compatibility Agarose Gels
Green Features Sustainable packaging
Product Line pUC19 DNA/MspI (HpaII) Marker
Quantity 50 μg