missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PdiI (NaeI) (10 U/μL)

Product Code. 10784681 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1,000 units
200 units
Unit Size:
1000 units
200 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10784681 200 units 200 units
10304630 1,000 units 1000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10784681 Supplier Thermo Scientific™ Supplier No. ER1521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PdiI (NaeI) restriction enzyme recognizes GCC^GGC sites and cuts best at 37°C in Tango buffer (Isoschizomers: MroNI, NaeI, NgoMIV).

pBR322 DNA digested with PdiI (NaeI), 0.7% agarose, 4 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G C C▵G G C...3'

3'...C G G▵C C G...5'

Conditions for 100% Activity:

  • 1X Buffer Tango: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • PdiI is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 500mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 0.15% Triton X-100 and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with PdiI, more than 90% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: never overlaps —no effect
  • CpG: completely overlaps —blocked
  • EcoKI: never overlaps —no effect
  • EcoBI: never overlaps —no effect

Digestion of Agarose-embedded DNA:

  • Minimum 20units of the enzyme are required for complete digestion of 1μg of agarose-embedded pBR322 DNA in 16hours

Note:

Certain sites in pBR322 are difficult to cleave with PdiI, the same as with its prototype NaeI

Assayed using pBR322 DNA (#SD0041)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme PdiI (NaeI)
Compatible Buffer 10X Buffer Tango
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 200 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.