Learn More
Description
pBR322 DNA digested with PdiI (NaeI), 0.7% agarose, 4 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...G C C▵G G C...3'
3'...C G G▵C C G...5'
Conditions for 100% Activity:
- 1X Buffer Tango: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- PdiI is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 500mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 0.15% Triton X-100 and 50% (v/v) glycerol
Ligation and Recleavage:
- After 50-fold overdigestion with PdiI, more than 90% of the DNA fragments can be ligated and recut
Methylation Effects:
- Dam: never overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: completely overlaps —blocked
- EcoKI: never overlaps —no effect
- EcoBI: never overlaps —no effect
Digestion of Agarose-embedded DNA:
- Minimum 20units of the enzyme are required for complete digestion of 1μg of agarose-embedded pBR322 DNA in 16hours
Note:
Certain sites in pBR322 are difficult to cleave with PdiI, the same as with its prototype NaeI
Assayed using pBR322 DNA (#SD0041)
Specifications
Specifications
| Concentration | 10 U/μL |
| Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
| Enzyme | PdiI (NaeI) |
| Compatible Buffer | 10X Buffer Tango |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | No |
| Quantity | 1,000 units |
| Product Type | Restriction Enzyme |
| Research Category | Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.