missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ O'GeneRuler Low Range DNA Ladder, Ready-to-Use 25-700 bp

Product Code. 10212840 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 Applications
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 10212840

Brand: Thermo Scientific™ SM1203

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform sizing and approximate quantification of double-stranded DNA on agarose gels with this Low range DNA ladder, premixed with blue and yellow tracking dyes.

O'GeneRuler Low Range DNA Ladder 25-700 bp, ready-to-use is recommended for sizing and approximate quantification of double-stranded DNA on agarose or polyacrylamide gels. It is ideal for denaturing polyacrylamide gel electrophoresis. The ladder is a mixture of chromatography-purified individual DNA fragments.

O'GeneRuler Low Range DNA Ladder, ready-to-use is premixed with 6X Orange DNA Loading Dye for direct loading on gel. 6X Orange DNA Loading Dye contains xylene cyanol FF and orange G. In a 1% agarose gel, orange G dye migrates at 50 bp. O'GeneRuler Low Range DNA Ladder is ideal when visualization of small DNA fragments is important.

The ladder is supplied with 6X Orange DNA Loading Dye for sample DNA.

Highlights

  • Ideal for both DNA sizing and approximate quantification
  • Ideal for non-denaturing polyacrylamide gel electrophoresis
  • Sharp bands
  • Bright reference bands (given in red)
  • Can be directly loaded and are stable at room temperature for 6 months
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Concentration 6X Loading Dye
Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Kit Contents 6X Orange DNA Loading Dye, 1.00mL
Quantity 100 Applications

This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.