missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ O'GeneRuler 1 kb DNA Ladder, Ready-to-Use- 250-10,000 bp

Product Code. 11551615 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 Applications
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11551615 500 Applications Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11551615 Supplier Thermo Scientific™ Supplier No. SM1163

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels with this 1 kb DNA ladder, premixed with blue and yellow tracking dyes.

O'GeneRuler 1kb DNA Ladder - 250-10,000 bp, ready-to-use is recommended for sizing and approximate quantification of a wide range of double-stranded DNA fragments in agarose gels. The ladder is a mixture of chromatography-purified individual DNA fragments.

O'GeneRuler 1kb DNA Ladder, ready-to-use is premixed with 6X Orange DNA Loading Dye, which contains xylene cyanol FF and orange G. In a 1% agarose gel, orange G dye migrates at 50bp. O'GeneRuler 1kb DNA Ladder is ideal when visualization of small DNA fragments is important.

The ladder is supplied with6X Orange DNA Loading Dye for sample DNA.

Highlights

  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Can be directly loaded and are stable at room temperature for 6months
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY

Specifications

Color Blue, Orange
Concentration 6X Loading Dye
Form Liquid
Kit Contents 6X Orange DNA Loading Dye, 2 x 1.00mL
Quantity 500 Applications
Product Type DNA Molecular Weight Marker

This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.