missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NotI, HC (50 U/μL)

Product Code. 10591031 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1,500 units
Unit Size:
1500 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10591031 1,500 units 1500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10591031 Supplier Thermo Scientific™ Supplier No. ER0593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Fermentas NotI is available as a FastDigest enzyme which is used for rapid DNA digestion.

Thermo Scientific™ NotI is available as a FastDigest enzyme which is used for rapid DNA digestion, more than 95% of the digested DNA fragments can be ligated and recut at a 5'-termini concentration of 0.5μM after a 50-fold overdigestion (3U/μg DNA x 17 hours) with NotI.

5'..G C▵G G C C G C...3'

3'..C G C C G G▵C G...5'

Conditions for 100% Activity:

  • 1X Buffer O: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • NotI is supplied in: 20mM Tris-HCl (pH 7.8 at 25°C), 100mM NaCl, 0.1mM EDTA, 1mM DTT, 0.02% Triton X-100, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with NotI, more than 95% of DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: never overlaps —no effect
  • CpG: completely overlaps —blocked
  • EcoKI: never overlaps —no effect
  • EcoBI: never overlaps —no effect

Digestion of Agarose-embedded DNA:

  • Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded Adenovirus-2 DNA in 16 hours

Compatible Ends:

  • Bsp120I, CfrI, Eco52I

Note:

Assayed using pTZ19RJL2 plasmid DNA digested with BseLI. Supercoiled plasmids may require up to 5-fold more NotI for complete digestion than linear DNAs.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 50 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Not I
Compatible Buffer 10X Buffer O
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 1,500 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.