missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ NBT
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
Thermo Scientific™ Fermentas NBT (nitro blue tetrazolium) is an oxidant used in conjunction with BCIP or BCIP derivatives (BCIP T), to enhance blue color development during detection of alkaline phosphatase activity.
It is suitable for immunohistochemistry and cytochemistry. NBT is also a substrate for oxidases and dehydrogenases.
Recommended for:
Detection of alkaline phosphatase activity in:
- Southern blotting
- Western blotting
- Northern blotting
- dot/slot blots
- plaque and colony screening
Note:
Preparation of a 75mg/mL stock solution in 70% dimethylformamide is recommended.
Specifications
Specifications
| For Use With (Application) | Detection of alkaline phosphatase activity in:; Southern blotting Western blotting Northern blotting dot/slot blots plaque and colony screening |
| Product Type | NBT |
| Quantity | 1 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction