missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MboII (5 U/μL)

Product Code. 10272450 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1,500 units
300 units
missing translation for 'unitSize'
1500 units
300 units
This item is not returnable. View return policy

Product Code. 10272450

missing translation for 'mfr': Thermo Scientific™ ER0821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MboII restriction enzyme recognizes GAAGA(8/7)^ sites and cuts best at 37°C in B buffer.

Lambda DNA (dam-) digested with MboII, 1.4% agarose, 130 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G A A G A (N)8▵...3'

3'...C T T C T (N)7▵...5'

Conditions for 100% Activity:

  • 1X Buffer B: 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2 and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • MboII is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Methylation Effects:

  • Dam: may overlap —blocked
  • Dcm: never overlaps —no effect
  • CpG: may overlap —no effect
  • EcoKI: never overlaps —no effect
  • EcoBI: may overlap —no effect

Note:

Assayed using lambda DNA (dam) (#SD0021). MboII is blocked by overlappingdam methylation. To avoiddam methylation, use adam,dcm- strain such as GM2163 (#M0099). Greater than 15-fold overdigestion with MboII may result in star activity. MboII produces DNA fragments that have a single-base 3 extension which are more difficult to ligate than blunt-ended fragments. MboII may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Methylation Sensitivity Dam Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme MboII
Compatible Buffer 10X Buffer B
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE Yes
Quantity 300 units
Product Type Restriction Enzyme
Research Category Scarless Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.