missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase

Product Code. 10394380 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 1.25mL
1000U
2 x 1.25mL
500U
Unit Size:
100 reactions
500 reactions
500 units
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10394380 500U 500 units
10763721 1000U Pack of 5
10140740 - -
11886833 2 x 1.25mL 100 reactions
11883923 10 x 1.25mL 500 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10394380 Supplier Thermo Scientific™ Supplier No. EP0602

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enhance the specificity, sensitivity, and yield of DNA amplification with this hot start DNA polymerase.

Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.

In addition, the enzyme provides the convenience of reaction set-up at room temperature. Maxima Hot Start Taq DNA Polymerase is a recombinant Taq DNA polymerase which has been chemically modified by the addition of heat-labile blocking groups to its amino acid residues. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification (see Fig. 1). The functional activity of the enzyme is restored during a short 4-minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase: catalyzes 5'=>3' synthesis of DNA, has no detectable 3'=>5' proofreading exonuclease activity, but possesses low 5'=>3' exonuclease activity. It exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Before activation, the two activities are not detectable.
  • High yield amplification of complex templates
  • Four minutes activation time
  • High PCR specificit—reduced effects of mispriming and primer-dimer formation
  • Enhanced PCR sensitivity
  • Convenient room temperature PCR set-up
  • Generates PCR products with 3'-dA overhangs

Recommended for:

Hot start PCR; High yield amplification of targets up to 3kb; RT-PCR.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 x 1.25mL of 10X Hot Start PCR Buffer, 2 x 1.25mL of 25mM MgCl2
Description Maxima™ Hot Start™ Taq DNA Polymerase
Quantity 500U
For Use With (Application) Cloning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.