missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MassRuler Express LR Forward and Reverse DNA Ladders

Product Code. 10736231 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 to 200 Applications
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 10736231

Brand: Thermo Scientific™ SM1273

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform fast and reliable DNA quantification in short separation distances under various electrophoresis conditions with these specially designed DNA Ladder mixes.

MassRuler Express LR Forward and Reverse DNA ladders are specially designed for fast and reliable DNA quantification in short separation distances under various electrophoresis conditions. In the forward versions, the mass of each DNA fragment is directly proportional to the fragment size, whereas in the reverse versions, the mass is inversely proportional to fragment's size.

The intensity of the fragment in each ladder is calibrated against a standard that guarantees the precise quantity of each band. The ladders are mixtures of chromatography-purified, individual DNA fragments.

Highlights

  • Accurate DNA sizing and quantification -DNA ladder fragments are quantified spectrophotometrically
  • Fast and reliable DNA quantification -short separation distances under various electrophoresis conditions
  • Sharp bands
  • Easy-to-remember fragment sizes and quantities
  • Ready-to-use -premixed with 6X MassRuler DNA Loading Dye for direct loading and room temperature storage
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY

Specifications

Concentration 6X Loading Dye
Product Type DNA Molecular Weight Marker/Ladder
Kit Contents 6X MassRuler™ DNA Loading Dye, 1mL
Quantity 50 to 200 Applications
Storage Requirements Room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.

This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.