missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LweI (SfaNI) (10 U/μL)

Product Code. 10389899 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 units
500 units
Unit Size:
100 units
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10389899 500 units 500 units
10252410 100 units 100 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10389899 Supplier Thermo Scientific™ Supplier No. ER1622

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The LweI (SfaNI) restriction enzyme recognizes GCATC(5/9)^ sites and cuts best at 37°C in Tango buffer (Isoschizomers: SfaNI).

Lambda DNA digested with LweI (SfaNI), 1.4% agarose, 169 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'..G C A T C (N)5▵...3'

3'..C G T A G (N)9▵...5'

Conditions for 100% Activity:

  • 1X Buffer Tango™: 33mM Tris-acetate (pH7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • LweI is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 100mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: never overlaps —no effect
  • CpG: may overlap —cleavage impaired
  • EcoKI: never overlaps —no effect
  • EcoBI: may overlap —effect not determined

Note:

LweI may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis. At least two copies of LweI recognition site are required for efficient cleavage.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme LweI (SfaNI)
Compatible Buffer 10X Buffer Tango
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE Yes
Quantity 500 units
Product Type Restriction Enzyme
Research Category Scarless Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.