missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Long PCR Enzyme Mix Not available in the USA

Product Code. 10460280 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100U
500U
Unit Size:
100 units
500 units
This item is not returnable. View return policy

Product Code. 10460280

Brand: Thermo Scientific™ K0181

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A unique enzyme blend for long and tricky PCR applications.

Thermo Scientific Long PCR Enzyme Mix is a unique blend of Taq DNA Polymerase and a thermostable high fidelity DNA polymerase with proofreading activity. The two enzymes synergistically generate long PCR products with greater yield and fidelity than Taq DNA Polymerase alone.

The fidelity of PCR with this enzyme mix is three times higher than with Taq DNA Polymerase. The ratio of enzymes in the Long PCR Enzyme Mix is optimized for generation of very long amplicons: up to 47 kb with viral DNA and up to 21 kb with genomic DNA templates. The specially formulated Long PCR Buffer protects DNA from depurination and nicking during long thermal cycling. The products generated with the Long PCR Enzyme Mix are mostly 3'-dA tailed. Long PCR Enzyme Mix is also used for efficient amplification of GC-rich DNA regions.

The two enzymes synergistically generate long PCR products with greater yield and fidelity than Taq DNA Polymerase alone. The fidelity of PCR with this enzyme mix is three times higher than with Taq DNA Polymerase. The ratio of enzymes in the Long PCR Enzyme Mix is optimized for generation of very long amplicons: up to 47kb with viral DNA and up to 21kb with genomic DNA templates (see Fig. 1). The specially formulated Long PCR Buffer protects DNA from depurination and nicking during long thermal cycling. The products generated with the Long PCR Enzyme Mix are mostly 3'-dA tailed.Long PCR Enzyme Mix is also used for efficient amplification of GC-rich DNA regions (see Fig. 2).
  • Unique blend of two thermostable DNA polymerases
  • Long PCR products:—up to 47kb with viral DNA as template,—up to 21kb with genomic DNA as template
  • Ideal for GC-rich templates up to 85%
  • Fidelity is three-times higher than with Taq DNA polymerase (see Fig. 1)
  • High yield

Recommended for:

Long range PCR; PCR from GC-rich or difficult templates; Generation of template for TA cloning; RT-PCR.

TRUSTED_SUSTAINABILITY

Specifications

Description Long PCR Enzyme Mix
Quantity 100U
For Use With (Application) Cloning

This product is licensed under one U.S. Patent No. 5,436,149 or corresponding foreign patents owned by Takara Bio, Inc.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.