missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LguI (SapI) (5 U/μL)

Product Code. 10660661 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 units
500 units
Unit Size:
100 units
500 units
This item is not returnable. View return policy

Product Code. 10660661

Brand: Thermo Scientific™ ER1932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The LguI (SapI) restriction enzyme recognizes GCTCTTC(1/4)^ sites and cuts best at 37°C in Tango buffer (Isoschizomers: BspQI, PciSI, SapI).

Lambda DNA digested with LguI (SapI), 0.7% agarose, 10 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'..G C T C T T C (N)1▵...3'

3'..C G A G A A G (N)4▵...5'

Conditions for 100% Activity:

  • 1X Buffer Tango™: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • LguI is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: never overlaps —no effect
  • CpG: may overlap —no effect
  • EcoKI: never overlaps —no effect
  • EcoBI: may overlap —effect not determined

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Note:

Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme LguI (SapI)
Compatible Buffer 10X Buffer Tango
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE Yes
Quantity 500 units
Product Type Restriction Enzyme
Research Category Scarless Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.