missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KpnI (10 U/μL)

Product Code. 10328759 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4,000 units
5 x 4,000 units
Unit Size:
4000 units
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10328759 4,000 units 4000 units
10338759 5 x 4,000 units Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10328759 Supplier Thermo Scientific™ Supplier No. ER0521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The KpnI restriction enzyme recognizes GGTAC^C sites and cuts best at 37°C in its own unique buffer (Isoschizomers: Asp718I).

Lambda DNA digested with KpnI, 0.7% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'..G G T A C▵C...3'

3'..C▵C A T G G...5'

Conditions for 100% Activity:

  • 1X Buffer KpnI: 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 0.02% Triton X-100 and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • KpnI is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: may overlap —no effect
  • CpG: may overlap —no effect
  • EcoKI: never overlaps —no effect
  • EcoBI: never overlaps —no effect

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Note:

High glycerol (>5%) concentrations, pH>8.0 or a large excess of enzyme may result in star activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Kpn I
Compatible Buffer Unique Buffer (10X Buffer KpnI)
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 4,000 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.