missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KpnI, HC (50 U/μL)

Product Code. 10785831 Shop All Thermo Scientific Products
Click to view available options
Quantity:
20,000 units
Unit Size:
20000 units
This item is not returnable. View return policy

Product Code. 10785831

Brand: Thermo Scientific™ ER0523

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Fermentas KpnI is available as a FastDigest enzyme which is used for rapid DNA digestion.

Thermo Scientific™ KpnI is available as a FastDigest enzyme which is used for rapid DNA digestion. More than 95% of the digested DNA fragments can be ligated and recut at a 5'-termini concentration of 0.02μM after a 50-fold overdigestion (3U/μg DNA x 17 hours) with KpnI.

5'..G G T A C▵C...3'

3'..C▵C A T G G...5'

Conditions for 100% Activity:

  • 1X Buffer KpnI: 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 0.02% Triton X-100 and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • KpnI is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: may overlap —no effect
  • CpG: may overlap —no effect
  • EcoKI: never overlaps —no effect
  • EcoBI: never overlaps —no effect

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Note:

High glycerol (>5%) concentrations, pH>8.0 or a large excess of enzyme may result in star activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 50 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Kpn I
Compatible Buffer Unique Buffer (10X Buffer KpnI)
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 20,000 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.