missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pfu DNA Polymerase

Product Code. 10631231 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Pfu DNA Polymerase
Pfu DNA Polymerase (recombinant)
Quantity:
100U
500U
Unit Size:
100 units
500 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity unitSize
10631231 Pfu DNA Polymerase 100U 100 units
10180690 Pfu DNA Polymerase 500U 500 units
10339879 Pfu DNA Polymerase (recombinant) 100U 100 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10631231 Supplier Thermo Scientific™ Supplier No. EP0571

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Optimize PCR applications requiring high fidelity with this highly thermostable DNA polymerase.

Pfu DNA Polymerase is a highly thermostable DNA polymerase from the hyperthermophilic archaeum Pyrococcus furiosus. The enzyme catalyzes the template-dependent polymerization of nucleotides into duplex DNA in the 5’→3’ direction. Pfu DNA Polymerase also exhibits 3’→5’ exonuclease (proofreading) activity that enables the polymerase to correct nucleotide incorporation errors. It has no 5’→3’ exonuclease activity.

Highlights

  • Eight times more accurate than Taq DNA polymerase
  • Highly thermostable – remains 95% active after 2 hours incubation at 95°C
  • Generates blunt-end PCR products
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescently-labeled nucleotides)
  • Increased PCR yields with native Pfu DNA Polymerase
TRUSTED_SUSTAINABILITY

Specifications

Description Pfu DNA Polymerase
Quantity 100U
For Use With (Application) High-fidelity PCR, Standard PCR, Cloning, RT-PCR, Site-directed Mutagenesis
Thermostability remains 95% active after 2 hours incubation at 95°C
Includes 0.6mL 10X Pfu Buffer with (NH4)2SO2, 0.60mL 10X Pfu Buffer with MgSO4 and 0.06mL 25mM MgSO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.