Learn More
Thermo Scientific™ HpaII (10 U/μL)
The HpaII restriction enzyme recognizes C^CGG sites and cuts best at 37°C in Tango buffer.
Brand: Thermo Scientific™ ER0512
Code : 88
Additional Details : Weight : 1.51950kg
Description
Lambda DNA digested with HpaII, 1.4% agarose, 328 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...C▵C G G...3'
3'...G G C▵C...5' Conditions for 100% Activity:
- 1X Buffer Tango™: 33mM Tris-acetate (pH7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- HpaII is supplied in: 10mM Tris-HCl (pH7.4 at 25°C), 100mM NaCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Methylation Effects:
- Dam: never overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: completely overlaps —blocked
- EcoKI: never overlaps —no effect
- EcoBI: never overlaps —no effect
Compatible Ends:
- Bsp119I, Bsu15I, Hin1I, MaeII, NarI, Psp1406I, SsiI, TaqI, XmiI
Specifications
10 U/μL | |
Hpa II | |
Yes | |
No | |
Restriction Enzyme |
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive | |
10X Buffer Tango | |
37°C | |
5000 U | |
Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.