Learn More
Thermo Scientific™ High Fidelity PCR Enzyme Mix Not available in US
Product Code. 10191370 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500U
Unit Size:
500 units
Product Code. Quantity unitSize
10191370 500U 500 units
1 options
Product Code. 10191370 Supplier Thermo Scientific™ Supplier No. K0192
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A Blend of Taq DNA Polymerase and a thermostable DNA polymerase with proofreading activity.
Thermo Scientific High Fidelity PCR Enzyme Mix is a unique blend of Taq DNA Polymerase and a thermostable DNA polymerase with proofreading activity. The two enzymes synergistically generate longer PCR products with greater yield and fidelity than Taq DNA Polymerase alone. Thus, fidelity of the High Fidelity PCR Enzyme Mix exceeds more than four times the fidelity of Taq DNA Polymerase. In addition, the enzyme mix can generate long amplicons up to 20 kb with viral DNA templates and up to 10 kb with genomic DNA templates. Such long amplicons are often difficult to generate with a single DNA polymerase.
The two enzymes synergistically generate longer PCR products with greater yield and fidelity than Taq DNA Polymerase alone. Thus, fidelity of the High Fidelity PCR Enzyme Mix exceeds more than four times the fidelity of Taq DNA Polymerase. In addition, the enzyme mix can generate long amplicons up to 20kb with viral DNA templates and up to 10kb with genomic DNA templates. Such long amplicons are often difficult to generate with a single DNA polymerase.
  • Unique blend of two thermostable DNA polymerases
  • Four-times greater accuracy than Taq DNA polymerase
  • Long amplification products:—up to 20kb with viral DNA as template, —up to 10kb with genomic DNA as template
  • High yields
  • Robust
Recommended for:
Generation of PCR products for purposes of cloning, expression, site-directed mutagenesis; High yield PCR; Long range PCR; RT-PCR.
Note:
High Fidelity PCR Enzyme Mix generates both blunt-ended and 3'-dA tailed PCR products. Blunting of PCR product prior to cloning is highly recommended. For efficient PCR product cloning, use the CloneJET PCR Cloning Kit.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500U
In certain countries use of this product is covered by patents. Purchase of product in these countries includes non-transferable, limited license for using only this amount of product for the purchaser's own internal research.
This product is licensed under one U.S. Patent No. 5,436,149 or corresponding foreign patents owned by Takara Bio, Inc.