missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FspAI (5 U/μL)

Product Code. 10304430 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 units
500 units
Unit Size:
100 units
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10304430 100 units 100 units
10620461 500 units 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10304430 Supplier Thermo Scientific™ Supplier No. ER1661

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The FspAI restriction enzyme recognizes RTGC^GCAY sites and cuts best at 37°C in O buffer.

Lambda DNA digested with FspAI, 0.5% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

More than 80% of the digested DNA fragments can be ligated and more than 90% can be recut at a 5ft.-termini concentration of 0.2μM after a 50-fold overdigestion (3U/μg DNA x 17 hours) with FspAI.

5'...R T G C▵G C A Y...3'

3'...Y A C G▵C G T R...5'

Conditions for 100% Activity:

  • 1X Buffer O: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • FspAI is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Effects:

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: completely overlaps — blocked
  • EcoKI: may overlap — effect not determined
  • EcoBI: may overlap — effect not determined

Note:

Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme FspAI
Compatible Buffer 10X Buffer O
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 100 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.