Learn More
Thermo Scientific™ Eco91I (BstEII) (10 U/μL)
The Eco91I (BstEII) restriction enzyme recognizes G^GTNACC sites and cuts best at 37°C in O buffer (Isoschizomers: BstEII, BstPI, EcoO65I, PspEI).
Brand: Thermo Scientific™ ER0391
27.32 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 88
Additional Details : Weight : 1.51950kg
Description
Lambda DNA digested with Eco91I (BstEII), 0.7% agarose, 13 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...G▵G T N A C C...3'
3'...C C A N T G▵G...5'
Conditions for 100% Activity:
- 1X Buffer O: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- Eco91I is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage:
- After 50-fold overdigestion with Eco91I, more than 95% of the DNA fragments can be ligated and recut
Methylation Effects:
- Dam: never overlaps-no effect
- Dcm: may overlap-no effect
- CpG: may overlap-no effect
- EcoKI: may overlap-effect not determined
- EcoBI: may overlap-effect not determined
Digestion of Agarose-embedded DNA:
- Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours
Compatible Ends:
- G▵GTSACC-MaeIII, NmuCI;G▵GTWACC-MaeIII
Specifications
10 U/μL | |
Eco91I (BstEII) | |
Yes | |
No | |
Restriction Enzyme |
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
10X Buffer O | |
37°C | |
1000 U | |
Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.