missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eco57I (AcuI) (5 U/μL)

Product Code. 10793711 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1,000 units
200 units
Unit Size:
1000 units
200 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10793711 200 units 200 units
10304910 1,000 units 1000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10793711 Supplier Thermo Scientific™ Supplier No. ER0341

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Eco57I (AcuI) restriction enzyme recognizes CTGAAG(16/14)^ sites and cuts best at 37°C in G (+SAM) buffer (Isoschizomers: AcuI).

Lambda DNA digested with Eco57I (AcuI), 1% agarose, 40 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...C T G A A G (N)16▵...3'

3'...G A C T T C (N)14▵...5'

Conditions for 100% Activity:

  • [1X Buffer G] + SAM:[10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 50mM NaCl and 0.1mg/mL BSA] + 0.01mM S-adenosylmethionine
  • Incubate at 37°C

Storage Buffer:

  • Eco57I is supplied in:10mM potassium phosphate (pH 7.4 at 25°C), 100mM NaCl, 1mM EDTA, 1mM DTT and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 10-fold overdigestion with Eco57I, approximately 70% of the DNA fragments can be ligated, but none of these can be recut due to the methylation of the recognition sequence by this enzyme

Methylation Effects:

  • Dam: never overlaps-no effect
  • Dcm: never overlaps-no effect
  • CpG: never overlaps-no effect
  • EcoKI: never overlaps-no effect
  • EcoBI: may overlap-effect not determined

Note:

Eco57I requires only Mg2 + for its activity, but is stimulated by S-adenosylmethionine. 0.01mM S-adenosylmethionine gives a 100-fold increase in Eco57I activity. Still, complete cleavage of some substrates with Eco57I is difficult to achieve. Eco57I concentration is determined by the maximum cleavage level achieved when no change in the fragmentation pattern is observed with further enzyme increase. Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity. Eco57I may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Eco57I (AcuI)
Compatible Buffer 10X Buffer G
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE Yes
Quantity 200 units
Product Type Restriction Enzyme
Research Category Scarless Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.