missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eco52I (EagI) (10 U/μL)

Product Code. 10389679 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2,500 units
500 units
Unit Size:
2500 units
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10389679 500 units 500 units
10319339 2,500 units 2500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10389679 Supplier Thermo Scientific™ Supplier No. ER0331

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The Eco52I (EagI) restriction enzyme recognizes C^GGCCG sites and cuts best at 37°C in its own unique buffer (Isoschizomers: BseX3I, BstZI, EagI, EclXI).

Lambda DNA digested with Eco52I (EagI), 0.7% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...C▵G G C C G...3'

3'...G C C G G▵C...5'

Conditions for 100% Activity:

  • 1X Buffer Eco52I:10mM Tris-HCl (pH 8.5 at 37°C), 3mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Eco52I is supplied in:10mM Tris-HCl (pH 8.2 at 25°C), 500mM NaCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with Eco52I, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps-no effect
  • Dcm: never overlaps-no effect
  • CpG: completely overlaps-blocked
  • EcoKI: never overlaps-no effect
  • EcoBI: never overlaps-no effect

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Compatible Ends:

  • Bsp120I, CfrI, NotI
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Eco52I (EagI)
Compatible Buffer Unique Buffer (10X Buffer Eco52I (EagI))
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 500 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.