missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eco47III (AfeI) (10 U/μL)

Product Code. 10223160 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1,000 units
200 units
Unit Size:
1000 units
200 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10223160 200 units 200 units
10690071 1,000 units 1000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10223160 Supplier Thermo Scientific™ Supplier No. ER0321

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The Eco47III (AfeI) restriction enzyme recognizes AGC^GCT sites and cuts best at 37°C in O buffer (Isoschizomers: AfeI, Aor51HI).

Lambda DNA digested with Eco47III (AfeI), 0.7% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...A G C▵G C T...3'

3'...T C G▵C G A...5'

Conditions for 100% Activity:

  • 1X Buffer O:50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Eco47III is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with Eco47III, more than 80% of pBR322 DNA fragments can be ligated and more than 90% of these can be recut

Methylation Effects:

  • Dam: never overlaps-no effect
  • Dcm: never overlaps-no effect
  • CpG: completely overlaps-blocked
  • EcoKI: never overlaps-no effect
  • EcoBI: may overlap-effect not determined

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Eco47III (AfeI)
Compatible Buffer 10X Buffer O
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 200 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.