missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eco47I (AvaII) (10 U/μL)

The Eco47I (AvaII) restriction enzyme recognizes G^GWCC sites and cuts best at 37°C in R buffer (Isoschizomers: AvaII, Bme18I, SinI, VpaK11BI).

Brand:  Thermo Scientific™ ER0312

Code : 88

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10839180

  • £164.00 / 4000 units
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Lambda DNA digested with Eco47I (AvaII), 1% agarose, 35 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G▵G W C C...3'

3'...C C W G▵G...5'

Conditions for 100% Activity:

  • 1X Buffer R:10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Eco47I is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with Eco47I, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps-no effect
  • Dcm: may overlap-blocked
  • CpG: may overlap-blocked
  • EcoKI: never overlaps-no effect
  • EcoBI: never overlaps-no effect

Compatible Ends:

  • Cfr13I, CpoI, Psp5II, SanDI

Note:

Eco47I is blocked by overlappingdcm methylation. To avoiddcm methylation, use adam,dcm- strain such as GM2163 (#M0099).

Specifications

Specifications

10 U/μL
37°C
4000 U
Eco47I (AvaII)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.