missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eco47I (AvaII) (10 U/μL)

Product Code. 10213160 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4000 U
800 units
Unit Size:
4000 units
800 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10213160 800 units 800 units
10839180 4000 U 4000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10213160 Supplier Thermo Scientific™ Supplier No. ER0311

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Eco47I (AvaII) restriction enzyme recognizes G^GWCC sites and cuts best at 37°C in R buffer (Isoschizomers: AvaII, Bme18I, SinI, VpaK11BI).

Lambda DNA digested with Eco47I (AvaII), 1% agarose, 35 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G▵G W C C...3'

3'...C C W G▵G...5'

Conditions for 100% Activity:

  • 1X Buffer R:10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Eco47I is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with Eco47I, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps-no effect
  • Dcm: may overlap-blocked
  • CpG: may overlap-blocked
  • EcoKI: never overlaps-no effect
  • EcoBI: never overlaps-no effect

Compatible Ends:

  • Cfr13I, CpoI, Psp5II, SanDI

Note:

Eco47I is blocked by overlappingdcm methylation. To avoiddcm methylation, use adam,dcm- strain such as GM2163 (#M0099).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
Enzyme Eco47I (AvaII)
Compatible Buffer 10X Buffer R
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 800 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.