Learn More
Thermo Scientific™ Eco147I (StuI) (10 U/μL)
The Eco147I (StuI) restriction enzyme recognizes AGG^CCT sites and cuts best at 37°C in B buffer (Isoschizomers: AatI, PceI, SseBI, StuI).
Brand: Thermo Scientific™ ER0421
Code : 88
Additional Details : Weight : 1.51950kg
Description
Lambda DNA digested with Eco147I (StuI), 0.7% agarose, 6 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...A G G▵C C T...3'
3'...T C C▵G G A...5'
Conditions for 100% Activity:
- 1X Buffer B:10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2 and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- Eco147I is supplied in:
- 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage:
- After 50-fold overdigestion with Eco147I, more than 90% of the DNA fragments can be ligated and recut
Methylation Effects:
- Dam: never overlaps — no effect
- Dcm: may overlap — blocked
- CpG: never overlaps — no effect
- EcoKI: never overlaps — no effect
- EcoBI: may overlap — effect not determined
Digestion of Agarose-embedded DNA:
- Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours
Note:
Eco147I is blocked by overlappingdcm methylation. To avoiddcm methylation, use adam,dcm- strain such as GM2163 (#M0099).
Specifications
10 U/μL | |
Eco147I (StuI) | |
Yes | |
No | |
Restriction Enzyme |
Dcm Methylation-Sensitive, Not Dam Methylation-Sensitive, Not CpG Methylation-Sensitive | |
10X Buffer B | |
37°C | |
1000 U | |
Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.