Learn More
Description
Lambda DNA digested with Eco105I (SnaBI), 0.7% agarose, 1 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...T A C▵G T A...3'
3'...A T G▵C A T...5'
Conditions for 100% Activity:
- 1X Buffer Tango™:
- 33mM Tris-acetate (pH7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA.Incubate at 37°C
Storage Buffer:
- Eco105I is supplied in:10mM Tris-HCl (pH7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 1mM phenylmethylsulfonylfluoride, 0.2mg/ml BSA and 50% (v/v) glycerol
Ligation and Recleavage:
- After 10-fold overdigestion with Eco105I, more than 80% of the M13mp18 DNA fragments can be ligated in a reaction mixture containing 20-40U of T4 DNA Ligase/1μg of fragments and 10% PEG
- More than 90% of these can be recut
Methylation Effects:
- Dam: never overlaps — no effect
- Dcm: never overlaps — no effect
- CpG: completely overlaps — blocked
- EcoKI: never overlaps — no effect
- EcoBI: never overlaps — no effect
Digestion of Agarose-embedded DNA:
- Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours
Note:
Greater than 15-fold overdigestion with Eco105I may result in star activity.
Specifications
Specifications
| Concentration | 10 U/μL |
| Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
| Enzyme | Eco105I (SnaBI) |
| Compatible Buffer | 10X Buffer Tango |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | No |
| Quantity | 600 units |
| Product Type | Restriction Enzyme |
| Research Category | Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.