Learn More
Thermo Scientific™ DecaLabel™ DNA Labeling Kit
Shop All Thermo Scientific ProductsDescription
Perform fast synthesis of radiolabeled DNA probes of high specific activity with the complete system provided by Thermo Scientific™ Fermentas DecaLabel™ DNA Labeling Kit.
Fast – 5 minutes at 37°C
High yields with Klenow Fragment, exo-: no degradation of a labeled probe during reaction
Flexible – suitable for either radiolabeled dATP or dCTP
Recommended for:
Generation of radiolabeled DNA probes for use in a variety of hybridization experiments: Southern and Northern blots, colony/plaque hybridizations, dot/slot blots and in situ hybridizations.
Specifications
Specifications
| For Use With (Application) | Generation of radiolabeled DNA probes for use in a variety of hybridization experiments: Southern and Northern blots, colony/plaque hybridizations, dot/slot blots and in situ hybridizations. |
| Includes | Klenow Fragment, exo-;Decanucleotides in Reaction Buffer;Mix A (-dATP);Mix C (-dCTP);dNTP Mix;Control Template;Water, nuclease-free;Detailed Protocol |
| No. of Reactions | 10 |
| Product Type | DecaLabel™ DNA Labeling Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.