missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BseXI (BbvI) (3 U/μL)

Product Code. 10359879 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 units
500 U
Unit Size:
100 units
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10359879 100 units 100 units
10714111 500 U 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10359879 Supplier Thermo Scientific™ Supplier No. ER1451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cut at GCAGC(8/12)^ sites with BseXI (BbvI) restriction enzyme, which performs best at 65°C in its own unique buffer (Isoschizomers: BbvI, BstV1I).

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G C A G C (N)8▵...3'

3'...C G T C G (N)12▵...5'

Conditions for 100% Activity:

  • 1X Buffer BseXI: 50mM Tris-HCl (pH 7.5 at 37°C), 2mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 65°C
  • To ensure higher efficiency of digestion, perform the cleavage reaction under paraffin oil

Storage Buffer:

  • BseXI is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 200mM NaCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 10-fold overdigestion with BseXI, more than 95% of the DNA fragments can be ligated recut

Methylation Effects:

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: may overlap — no effect
  • EcoKI: never overlaps — no effect
  • EcoBI: may overlap — effect not determined

Note:

Assayed using pBR322 DNA (#SD0041). Incubation at 37°C results in 10% activity. Greater than 40-fold overdigestion with BseXI may result in star activity. BseXI may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 3 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme BseXI (BbvI)
Compatible Buffer Unique Buffer (10X Buffer BseXI (BbvI))
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 65°C
Type IIS RE Yes
Quantity 100 units
Product Type Restriction Enzyme
Research Category Scarless Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.