missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BseDI (BsaJI) (10 U/μL)

Product Code. p-4521400 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1500 U
300 units
Unit Size:
1500 units
300 units
This item is not returnable. View return policy

Product Code. 10689330

Brand: Thermo Scientific™ ER1081

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cut at C^CNNGG sites with BseDI (BsaJI) restriction enzyme, which performs best at 55°C in Tango buffer (Isoschizomers: BsaJI, BssECI).

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...C▵C N N G G...3'

3'...G G N N C▵C...5'

Conditions for 100% Activity:

  • 1X Buffer Tango™: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 55°C

Storage Buffer:

  • BseDI is supplied in:10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with BseDI, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlap — no effect
  • Dcm: may overlap — no effect
  • CpG: may overlap — no effect
  • EcoKI: never overlaps — no effect
  • EcoBI: never overlaps — no effect

Note:

Incubation at 37°C results in 10% activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme BseDI (BsaJI)
Compatible Buffer 10X Buffer Tango
Sensitive to Heat Inactivation No
Optimal Reaction Temperature 55°C
Type IIS RE No
Quantity 300 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.