missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BseDI (BsaJI) (10 U/μL)

Product Code. 10374630 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1500 U
300 units
Unit Size:
1500 units
300 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10374630 1500 U 1500 units
10689330 300 units 300 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10374630 Supplier Thermo Scientific™ Supplier No. ER1082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cut at C^CNNGG sites with BseDI (BsaJI) restriction enzyme, which performs best at 55°C in Tango buffer (Isoschizomers: BsaJI, BssECI).

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...C▵C N N G G...3'

3'...G G N N C▵C...5'

Conditions for 100% Activity:

  • 1X Buffer Tango™: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 55°C

Storage Buffer:

  • BseDI is supplied in:10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with BseDI, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlap — no effect
  • Dcm: may overlap — no effect
  • CpG: may overlap — no effect
  • EcoKI: never overlaps — no effect
  • EcoBI: never overlaps — no effect

Note:

Incubation at 37°C results in 10% activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Incubator Temperature 55°C
Quantity 1500 U
Product Type BseDI (BsaJI)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.