missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Aminoallyl-dUTP Solution (50 mM)

Product Code. 10775641 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10775641 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10775641 Supplier Thermo Scientific™ Supplier No. R1101
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH.

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH. Aminoallyl-dUTP can be enzymatically incorporated into DNA and the resulting amine-containing DNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin or hapten.

Applications
• Enzymatic indirect non-radioactive labeling of DNA during cDNA synthesis
• PCR
• Nick-translation
• Random-primed labeling
• Primer extension

Aminoallyl-dUTP can be enzymatically incorporated into DNA with:
• Reverse transcriptases
• Taq DNA polymerase (Cat. No. EP0072)
• phi29 DNA Polymerase (EP0091)
• Klenow Fragment (EP0051)
• Klenow Fragment, exo- (EP0421)
• DNA Polymerase I (EP0041)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 50 mM
Content And Storage Aminoallyl-dUTP Solution, 50 mM (50 μL)

Store at –20°C.
Form Liquid
Polymerase DNA Polymerase
Labeling Method Direct Labeling
Label or Dye Aminoallyl
Quantity 50 μL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR, Gene Expression, Non-Radioactive Labeling

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.