missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FERD3L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541232
This item is not returnable.
View return policy
Description
Peptide sequence: SCVDTTVLDF VADLSLASPR RPLLCDFAPG VSLGDPALAL REGRPRRMAR Sequence homology: Human: 100%.
FERD3L is an evolutionarily conserved bHLH transcription factor that is identified in fly, mouse and man. Nervous system expression of this protein is detected in fly and mouse.
Specifications
| FERD3L | |
| Polyclonal | |
| Unconjugated | |
| FERD3L | |
| Basic helix-loop-helix protein N-twist; bHLHa31; Class A basic helix-loop-helix protein 31; fer3; Fer3-like (Drosophila); Fer3-like bHLH transcription factor; fer3-like protein; FERD3L; Mnato3; Nato; NATO3; nephew of atonal 3; Neuronal twist; NTWIST; N-TWIST | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 222894 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96RJ6 | |
| FERD3L | |
| Synthetic peptide directed towards the N-terminal of human FERD3L (aa 7-56). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction