missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FERD3L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | FERD3L |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
FERD3L Polyclonal specifically detects FERD3L in Human samples. It is validated for Western Blot.Specifications
| FERD3L | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Basic helix-loop-helix protein N-twist, BHLHA31, bHLHa31NTWIST, Class A basic helix-loop-helix protein 31, Fer3-like (Drosophila), fer3-like protein, MGC119861, NATO3Neuronal twist, Nephew of atonal 3, N-TWIST | |
| FERD3L | |
| IgG | |
| 19 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_690862 | |
| 222894 | |
| Synthetic peptide directed towards the N terminal of human FERD3L. Peptide sequence SCVDTTVLDFVADLSLASPRRPLLCDFAPGVSLGDPALALREGRPRRMAR. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title