missing translation for 'onlineSavingsMsg'
Learn More

Ferbam, TRC

Product Code. 30401047
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401047 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401047 Supplier Toronto Research Chemicals Supplier No. TRCF309508500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ferbam
CAS 14484-64-1
Molecular Formula C9 H18 Fe N3 S6
Quantity 500 mg
Synonym Iron, tris(N,N-dimethylcarbamodithioato-κS,κS')-, (OC-6-11)- (ACI), (OC-6-11)-Tris(N,N-dimethylcarbamodithioato-κS,κS')iron (ACI), Carbamodithioic acid, dimethyl-, iron complex (ZCI), Iron, tris(dimethylcarbamodithioato-S,S')-, (OC-6-11)- (ZCI), Iron, tris(dimethylcarbamodithioato-κS,κS')-, (OC-6-11)- (9CI), Iron, tris(dimethyldithiocarbamato)- (8CI), Aafertis, Bercema Fertam 50, Dimethyldithiocarbamic acid iron(3+) salt, Ferbam, Ferbam 50, Ferberk, Fermate, Ferradow, Ferric dimethyldithiocarbamate, Ferric N,N-dimethyldithiocarbamate, Fuklasin Ultra, Fuklazin, Hexaferb, Iron dimethyldithiocarbamate, Iron tris(dimethyldithiocarbamate), Karbam Black, Liromate, Nocceler TTFE, Sanceler TT-FE, Stauffer ferbam, Trimanzone, Tris(dimethyldithiocarbamato)iron, Tris(N,N-dimethyldithiocarbamato)iron, Tris(N,N-dimethyldithiocarbamato)iron(III)
InChI Formula InChI=1S/3C3H7NS2.Fe/c3*1-4(2)3(5)6;/h3*1-2H3,(H,5,6);/q;;;+3/p-3
SMILES CN(C)C(=S)S[Fe](SC(=S)N(C)C)SC(=S)N(C)C
IUPAC Name tris(dimethylcarbamothioylsulfanyl)iron
Molecular Weight (g/mol) 416.49
Formula Weight 415.9174
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.