missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fer Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-20089H
This item is not returnable.
View return policy
Description
Fer Polyclonal specifically detects Fer in Human samples. It is validated for Western Blot, Knockdown Validated.
Specifications
| Fer | |
| Polyclonal | |
| Western Blot, Knockdown Validated | |
| P16591 | |
| FER | |
| Synthetic peptide made to an internal portion of human FER (within residues 400-450). [Swiss-Prot# P16591] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Predicted to react with mouse and canine based on 100% sequence homology. | |
| Store at 4C in the dark. |
| Western Blot, KnockDown | |
| HRP | |
| PBS with No Preservative | |
| EC 2.7.10, EC 2.7.10.2, fer (fps/fes related) tyrosine kinase, p94-Fer, Proto-oncogene c-Fer, TYK3phosphoprotein NCP94, Tyrosine kinase 3, tyrosine-protein kinase Fer | |
| Rabbit | |
| 94 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 2241 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction