missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fer-1-like protein 6 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05936AF700
This item is not returnable.
View return policy
Description
Fer-1-like protein 6 Polyclonal specifically detects Fer-1-like protein 6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Fer-1-like protein 6 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C8ORFK23, FER1L6, Fer-1-Like 6 (C. Elegans) | |
| Produced in rabbits immunized with E. coli-derived Human Fer-1-like protein 6 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 654463 | |
| Store at 4°C in the dark. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido