missing translation for 'onlineSavingsMsg'
Learn More

Fenticonazole Sulfone Nitric Acid Salt, TRC

Product Code. 30136300
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136300 50 mg 50mg
17943005 10 mg 10mg
17942375 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136300 Supplier Toronto Research Chemicals Supplier No. TRCF27926050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80676-29-5
Molecular Formula C24 H20 Cl2 N2 O3 S . H N O3
Molecular Weight (g/mol) 550.41
Synonym 1-[2-(2,4-Dichlorophenyl)-2-[[4-(phenylsulfonyl)phenyl]methoxy]ethyl]-1H-imidazole Nitrate, Fenticonazole Sulfone Nitrate, 1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[4-(benzenesulfonyl)benzyl]oxy]ethyl]-1H-imidazole Nitrate, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[[4-(phenylsulfonyl)phenyl]methoxy]ethyl]-, nitrate (1:1), 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[[4-(phenylsulfonyl)phenyl]methoxy]ethyl]-, mononitrate (9CI)
IUPAC Name 1-[2-[[4-(benzenesulfonyl)phenyl]methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
SMILES O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3ccc(cc3)S(=O)(=O)c4ccccc4)c(Cl)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H20Cl2N2O3S.HNO3/c25-19-8-11-22(23(26)14-19)24(15-28-13-12-27-17-28)31-16-18-6-9-21(10-7-18)32(29,30)20-4-2-1-3-5-20;2-1(3)4/h1-14,17,24H,15-16H2;(H,2,3,4)
Formula Weight 549.0528
Chemical Name or Material Fenticonazole Sulfone Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.