missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenthoxon Sulfoxide, TRC
Specifications
Specifications
| Chemical Name or Material | Fenthoxon Sulfoxide |
| CAS | 6552-13-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H15 O5 P S |
| Quantity | 100 mg |
| Synonym | Fenthion Oxon Sulfoxide, Phosphoric acid, dimethyl 3-methyl-4-(methylsulfinyl)phenyl ester, Phosphoric acid, dimethyl 4-(methylsulfinyl)-m-tolyl ester (7CI,8CI), Fenoxon sulfoxide, Fenthion oxon sulfoxide, Fenthion-oxon-sulfoxide |
| InChI Formula | InChI=1S/C10H15O5PS/c1-8-7-9(5-6-10(8)17(4)12)15-16(11,13-2)14-3/h5-7H,1-4H3 |
| SMILES | COP(=O)(OC)Oc1ccc(c(C)c1)S(=O)C |
| IUPAC Name | dimethyl (3-methyl-4-methylsulfinylphenyl) phosphate |
| Molecular Weight (g/mol) | 278.2619 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?