missing translation for 'onlineSavingsMsg'
Learn More

Fenthion, TRC

Product Code. 30134513
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134513 10 g 10g
30134386 25 g 25g
17927979 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134513 Supplier Toronto Research Chemicals Supplier No. TRCF27800010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 55-38-9
Molecular Formula C10 H15 O3 P S2
Molecular Weight (g/mol) 278.33
Synonym Phosphorothioic acid, O,O-dimethyl O-[3-methyl-4-(methylthio)phenyl] ester (9CI, ACI), m-Cresol, 4-(methylthio)-, O-ester with O,O-di-Me phosphorothioate (6CI), Phosphorothioic acid (H3PO3S), O,O-dimethyl O-4-(methylthio)-m-tolyl ester (6CI), Phosphorothioic acid, dimethyl [4-(methylthio)-m-tolyl] ester (7CI), Phosphorothioic acid, O,O-dimethyl O-[4-(methylthio)-m-tolyl] ester (8CI), B 29493, BAY 29493, Baycid, Bayer 9007, Baytex, Dimethyl 4-methylthio-m-tolyl phosphorothioate, ENT 25540, Entex, Fenthion, Fenthion-methyl, Lebaycid, Mercaptofos, Mercaptophos, MeSH ID: D005284, MPP, MPP (pesticide), O,O-Dimethyl O-(3-methyl-4-methylmercaptophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-methylthiophenyl) thiophosphate, O,O-Dimethyl O-(4-methylthio-3-methylphenyl) phosphorothioate, O,O-Dimethyl O-4-(methylmercapto)-3-methylphenyl phosphorothioate, O,O-Dimethyl O-4-(methylmercapto)-3-methylphenyl thiophosphate, O,O-Dimethyl O-[3-methyl-4-(methylthio)phenyl] phosphorothioate, O,O-Dimethyl O-[4-(methylthio)-m-tolyl] phosphorothioate, O,O-Dimethyl O-[4-(methylthio)-m-tolyl] thiophosphate, OMS 2, Phenthion, Queletox, S 1752, Spotton, Sulfidophos, Talodex, Tiguvon
IUPAC Name dimethoxy-(3-methyl-4-methylsulfanylphenoxy)-sulfanylidene-λ^{5}-phosphane
SMILES COP(=S)(OC)Oc1ccc(SC)c(C)c1

Spezifikation

Quantity 10 g
InChI Formula InChI=1S/C10H15O3PS2/c1-8-7-9(5-6-10(8)16-4)13-14(15,11-2)12-3/h5-7H,1-4H3
Formula Weight 278.02
Chemical Name or Material Fenthion
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.