missing translation for 'onlineSavingsMsg'
Learn More

Fenthion Sulfone, TRC

Product Code. 18011630
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18011630 250 mg 250mg
18061520 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18011630 Supplier Toronto Research Chemicals Supplier No. TRCF278015250MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3761-42-0
Molecular Formula C10 H15 O5 P S2
Molecular Weight (g/mol) 310.33
Synonym Fenthion Sulfone, Phosphorothioic acid, O,O-dimethyl O-[3-methyl-4-(methylsulfonyl)phenyl] ester (9CI, ACI), Phosphorothioic acid, O,O-dimethyl O-[4-(methylsulfonyl)-m-tolyl] ester (7CI, 8CI), Fenthion sulfone
IUPAC Name dimethoxy-(3-methyl-4-methylsulfonylphenoxy)-sulfanylidene-λ^{5}-phosphane
SMILES COP(=S)(OC)Oc1ccc(c(C)c1)S(=O)(=O)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H15O5PS2/c1-8-7-9(15-16(17,13-2)14-3)5-6-10(8)18(4,11)12/h5-7H,1-4H3
Formula Weight 310.0099
Percent Purity >95%
Chemical Name or Material Fenthion-sulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.