missing translation for 'onlineSavingsMsg'
Learn More

Fensulfothion, TRC

Product Code. 30403188
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403188 500 mg
30402133 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30403188 Supplier Toronto Research Chemicals Supplier No. TRCF265490500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-90-2
Molecular Formula C11 H17 O4 P S2
Molecular Weight (g/mol) 308.35
Synonym Phosphorothioic acid, O,O-diethyl O-[4-(methylsulfinyl)phenyl] ester, Phosphorothioic acid, O,O-diethyl O-[p-(methylsulfinyl)phenyl] ester (6CI,7CI,8CI), Phenol, p-(methylsulfinyl)-, O-ester with O,O-di-Et phosphorothioate (6CI), Agricur, B 25141, BAY 25141, Chemagro 25141, DMSP, Daconit, Dasanit, ENT 24,945, Fensulfothion, O,O-Diethyl O-(4-methylsulfinylphenyl) monothiophosphate, O,O-Diethyl O-[4-(methylsulfinyl)phenyl] phosphorothioate, O,O-Diethyl O-[p-(methylsulfinyl)phenyl] phosphorothioate, S 767, Terracur P, VUAgt 108, VUAgt 96
IUPAC Name diethoxy-(4-methylsulfinylphenoxy)-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)Oc1ccc(cc1)S(=O)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H17O4PS2/c1-4-13-16(17,14-5-2)15-10-6-8-11(9-7-10)18(3)12/h6-9H,4-5H2,1-3H3
Formula Weight 308.0306
Chemical Name or Material Fensulfothion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.