missing translation for 'onlineSavingsMsg'
Learn More

Fensulfothion Sulfone, TRC

Product Code. 30041382
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041382 50 mg 50mg
30010661 500 mg 500mg
30012168 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041382 Supplier Toronto Research Chemicals Supplier No. TRCF26552050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14255-72-2
Molecular Formula C11 H17 O5 P S2
Molecular Weight (g/mol) 324.35
Synonym Phosphorothioic acid, O,O-diethyl O-[4-(methylsulfonyl)phenyl] ester, Phosphorothioic acid, O,O-diethyl O-[p-(methylsulfonyl)phenyl] ester (7CI,8CI), Phosphorothioic acid, diethyl O-[p-(methylsulfonyl)phenyl] ester (6CI), Phenol, p-(methylsulfonyl)-, O-ester with O,O-diethyl phosphorothioate (8CI), Dasanit sulfone, Dasanit sulphone, Fensulfothion sulfone, O,O-Diethyl O-(4-methylsulfonylphenyl) phosphorothioate
IUPAC Name diethoxy-(4-methylsulfonylphenoxy)-sulfanylidene-λ^{5}-phosphane
SMILES CCOP(=S)(OCC)Oc1ccc(cc1)S(=O)(=O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H17O5PS2/c1-4-14-17(18,15-5-2)16-10-6-8-11(9-7-10)19(3,12)13/h6-9H,4-5H2,1-3H3
Formula Weight 324.0255
Percent Purity >95%
Chemical Name or Material Fensulfothion-sulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.