missing translation for 'onlineSavingsMsg'
Learn More

Fensulfothion Oxon, TRC

Product Code. 30008375
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008375 500 mg 500mg
30009892 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008375 Supplier Toronto Research Chemicals Supplier No. TRCF265500500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6552-21-2
Molecular Formula C11 H17 O5 P S
Molecular Weight (g/mol) 292.29
Synonym Phosphoric acid, diethyl 4-(methylsulfinyl)phenyl ester, Phosphoric acid, diethyl p-(methylsulfinyl)phenyl ester (6CI,7CI,8CI), Phenol, p-(methylsulfinyl)-, diethyl phosphate (8CI), Dasanit O, Dasanit O analog, Diethyl 4-methylsulfinylphenyl phosphate, Fensulfothion oxon, NSC 133045
IUPAC Name diethyl (4-methylsulfinylphenyl) phosphate
SMILES CCOP(=O)(OCC)Oc1ccc(cc1)S(=O)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H17O5PS/c1-4-14-17(12,15-5-2)16-10-6-8-11(9-7-10)18(3)13/h6-9H,4-5H2,1-3H3
Formula Weight 292.0534
Percent Purity >95%
Chemical Name or Material Fensulfothion-oxon
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.