missing translation for 'onlineSavingsMsg'
Learn More

Fenspiride N-Oxide, TRC

Product Code. 30137602
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137602 100 mg 100mg
30012341 25 mg 25mg
30011462 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137602 Supplier Toronto Research Chemicals Supplier No. TRCF265010100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210690-26-9
Molecular Formula C15H20N2O3
Molecular Weight (g/mol) 276.33
Synonym trans- 8-(2-Phenylethyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one 8-Oxide, (5α,8β)-8-(2-Phenylethyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one 8-Oxide;
SMILES [O-][N+]1(CCc2ccccc2)CCC3(CC1)CNC(=O)O3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H20N2O3/c18-14-16-12-15(20-14)7-10-17(19,11-8-15)9-6-13-4-2-1-3-5-13/h1-5H,6-12H2,(H,16,18)
Chemical Name or Material Fenspiride N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.