missing translation for 'onlineSavingsMsg'
Learn More

Fenspiride Hydrochloride, TRC

Product Code. 30138674
Change view
Click to view available options
Quantity:
50 mg
1 g
2.5 g
Unit Size:
1g
2.5g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138674 2.5 g 2.5g
17944465 50 mg 50mg
17984655 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138674 Supplier Toronto Research Chemicals Supplier No. TRCF2650002.5G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5053-08-7
Molecular Formula C15 H20 N2 O2 . Cl H
Molecular Weight (g/mol) 296.79
Synonym 1-Oxa-3,8-diazaspiro[4.5]decan-2-one, 8-(2-phenylethyl)-, hydrochloride (1:1), 1-Oxa-3,8-diazaspiro[4.5]decan-2-one, 8-(2-phenylethyl)-, monohydrochloride (9CI), 1-Oxa-3,8-diazaspiro[4.5]decan-2-one, 8-phenethyl-, monohydrochloride (8CI), 8-Phenethyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one hydrochloride, Decaspir, Decaspiride, Erespal, Fenspiride hydrochloride, Fluiden, JP 428, NAT 333, NDR 5998A, Pneumorel, Tegencia, Viarespan
IUPAC Name 8-(2-phenylethyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one;hydrochloride
SMILES Cl.O=C1NCC2(CCN(CCc3ccccc3)CC2)O1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C15H20N2O2.ClH/c18-14-16-12-15(19-14)7-10-17(11-8-15)9-6-13-4-2-1-3-5-13;/h1-5H,6-12H2,(H,16,18);1H
Formula Weight 296.1292
Chemical Name or Material Fenspiride Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.