missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FENS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145224
This item is not returnable.
View return policy
Description
Immunogen sequence: VRVCDSCYDS IKDEDRTSLA TFHEGKHNIS HMSMDIARGL MVTCGTDRIV KI Antigen sequence identity: Mouse 98%; Rat 98%.
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains a single FYVE domain and multiple WD40 repeats. This protein is localized to endosomes.
Specifications
| FENS1 | |
| Polyclonal | |
| Unconjugated | |
| WDFY1 | |
| 1700013B03Rik; 1700120F24Rik; FENS1; FENS-1; FYVE domain-containing protein localized to endosomes 1; Jr1; KIAA1435; MGC137087 protein; mKIAA1435; Phosphoinositide-binding protein 1; phosphoinositide-binding protein SR1; WD repeat and FYVE domain containing 1; WD repeat and FYVE domain-containing protein 1; WD40 and FYVE domain containing 1; WD40- and FYVE domain-containing protein 1; WDF1; Wdfy1; ZFYVE17; zgc:101764; zgc:101764 protein; zgc:109909; zinc finger FYVE domain-containing protein 17 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57590 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IWB7 | |
| WDFY1 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 341-392. Recombinant protein control fragment (Product #RP-110069). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction