missing translation for 'onlineSavingsMsg'
Learn More

Fenquizone, TRC

Product Code. 30135710
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135710 50 mg 50mg
30007792 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135710 Supplier Toronto Research Chemicals Supplier No. TRCF24985050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20287-37-0
Molecular Formula C14 H12 Cl N3 O3 S
Molecular Weight (g/mol) 337.78
Synonym 7-Chloro-1,2,3,4-tetrahydro-4-oxo-2-phenyl-6-quinazolinesulfonamide;
IUPAC Name 1-(3-Bromo-4-fluorophenyl)propan-1-one
SMILES NS(=O)(=O)c1cc2C(=O)NC(Nc2cc1Cl)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H8BrFO/c1-2-9(12)6-3-4-8(11)7(10)5-6/h3-5H,2H2,1H3
Formula Weight 229.97
Chemical Name or Material Fenquizone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.